Product Details

SNP ID
rs11611207
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:104809478 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCTTTGGACATCTGTCAGAGAGAG[G/T]TTGTACTTTAAGCCCTATTGTCTTT
Phenotype
MIM: 610802
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC41A2 PubMed Links

Gene Details

Gene
SLC41A2
Gene Name
solute carrier family 41 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032148.3 Intron NP_115524.3
XM_005269176.1 Intron XP_005269233.1
XM_005269178.1 Intron XP_005269235.1
XM_005269179.1 Intron XP_005269236.1
XM_006719630.2 Intron XP_006719693.1
XM_011538807.2 Intron XP_011537109.1
XM_011538808.1 Intron XP_011537110.1
XM_011538809.1 Intron XP_011537111.1
XM_011538810.2 Intron XP_011537112.1
XM_011538811.2 Intron XP_011537113.1
XM_011538813.2 Intron XP_011537115.1
XM_017020013.1 Intron XP_016875502.1
XM_017020014.1 Intron XP_016875503.1
XM_017020015.1 Intron XP_016875504.1
XM_017020016.1 Intron XP_016875505.1
XM_017020017.1 Intron XP_016875506.1

View Full Product Details