Product Details

SNP ID
rs1474945
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:31258599 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGAGTTGGCTTGAGCATGCCTTAT[A/G]TTCCAGCGAGGGCAAGTAGAATAGA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DEFB115 PubMed Links
Additional Information
For this assay, SNP(s) [rs6120961] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DEFB115
Gene Name
defensin beta 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037730.1 Intron NP_001032819.1

View Full Product Details