Product Details

SNP ID
rs12433900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92927958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCACTGATGATATTCAAGCCAA[A/G]CCTGGACAGGATACTGGAGTGGAGG
Phenotype
MIM: 118910 MIM: 601838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHGA PubMed Links

Gene Details

Gene
CHGA
Gene Name
chromogranin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001275.3 Intron NP_001266.1
NM_001301690.1 Intron NP_001288619.1
XM_011536370.1 Intron XP_011534672.1
Gene
ITPK1
Gene Name
inositol-tetrakisphosphate 1-kinase
There are no transcripts associated with this gene.

View Full Product Details