Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286116.1 | 3681 | UTR 3 | NP_001273045.1 | ||
NM_001286117.1 | 3681 | UTR 3 | NP_001273046.1 | ||
NM_001286118.1 | 3681 | UTR 3 | NP_001273047.1 | ||
NM_001286119.1 | 3681 | UTR 3 | NP_001273048.1 | ||
NM_001286120.1 | 3681 | UTR 3 | NP_001273049.1 | ||
NM_021825.4 | 3681 | UTR 3 | NP_068597.2 | ||
XM_005274154.2 | 3681 | Intron | XP_005274211.1 | ||
XM_005274156.3 | 3681 | Intron | XP_005274213.1 | ||
XM_005274159.3 | 3681 | Intron | XP_005274216.1 | ||
XM_017018103.1 | 3681 | Intron | XP_016873592.1 | ||
XM_017018104.1 | 3681 | Intron | XP_016873593.1 |