Product Details

SNP ID
rs12455822
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:35335959 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTATCGTGGACTCACCTCTGAACAC[C/T]GGAGAGCCAGCTAGCTCTTTTGAGC
Phenotype
MIM: 194534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF24 PubMed Links

Gene Details

Gene
ZNF24
Gene Name
zinc finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308123.1 2558 Intron NP_001295052.1
NM_006965.3 2558 UTR 3 NP_008896.2
XM_005258341.3 2558 UTR 3 XP_005258398.1

View Full Product Details