Product Details

SNP ID
rs10934101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:111542584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATTTCCTTTCTAACAGGATCTC[C/T]GGAGTAATTGGCAAATTGTGTTTCT
Phenotype
MIM: 606037
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD96 PubMed Links

Gene Details

Gene
CD96
Gene Name
CD96 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318889.1 Intron NP_001305818.1
NM_005816.4 Intron NP_005807.1
NM_198196.2 Intron NP_937839.1
XM_005247063.3 Intron XP_005247120.1
XM_006713469.3 Intron XP_006713532.1
XM_006713470.3 Intron XP_006713533.1
XM_017005521.1 Intron XP_016861010.1
XM_017005522.1 Intron XP_016861011.1

View Full Product Details