Product Details

SNP ID
rs2445727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94373368 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCCAGGGCACTTAGTCACCTTT[C/T]CCAGCCTCCCTTACGGTTAGATATT
Phenotype
MIM: 604289
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAD54B PubMed Links

Gene Details

Gene
RAD54B
Gene Name
RAD54 homolog B (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205262.2 Intron NP_001192191.1
NM_001205263.1 Intron NP_001192192.1
NM_012415.3 Intron NP_036547.1

View Full Product Details