Product Details
- SNP ID
-
rs2399789
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:12198100 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TATCCCATAAGTTTTTAAGAGACTT[C/G]AGGTATGGCTTTGCCATCCTTAATT
- Phenotype
-
MIM: 609230
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDC123
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117274893,rs76448698] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDC123
- Gene Name
- cell division cycle 123
- Gene
- NUDT5
- Gene Name
- nudix hydrolase 5
There are no transcripts associated with this gene.
View Full Product Details