Product Details

SNP ID
rs10495148
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:220547065 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGATAAGCTAGTTTATCTTGGATC[A/C]AATCACTAACCTTTCTTTCTTATGG
Phenotype
MIM: 606511
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
MARK1 PubMed Links

Gene Details

Gene
MARK1
Gene Name
microtubule affinity regulating kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286124.1 Intron NP_001273053.1
NM_001286126.1 Intron NP_001273055.1
NM_001286128.1 Intron NP_001273057.1
NM_001286129.1 Intron NP_001273058.1
NM_018650.4 Intron NP_061120.3
XM_005273134.4 Intron XP_005273191.1
XM_006711326.3 Intron XP_006711389.1
XM_011509561.2 Intron XP_011507863.1
XM_011509562.2 Intron XP_011507864.1
XM_017001305.1 Intron XP_016856794.1
XM_017001306.1 Intron XP_016856795.1
XM_017001307.1 Intron XP_016856796.1

View Full Product Details