Product Details

SNP ID
rs945393
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:16043984 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCAGCACCTCACTTTGGCCCTGG[C/T]GGGATCTTCTTCTGGGCCTGGGGTT
Phenotype
MIM: 602024 MIM: 602023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCNKA PubMed Links

Gene Details

Gene
CLCNKA
Gene Name
chloride voltage-gated channel Ka
There are no transcripts associated with this gene.

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000085.4 Intron NP_000076.2
NM_001165945.2 Intron NP_001159417.2

View Full Product Details