Product Details
- SNP ID
-
rs11603815
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:108673118 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTGTCCTTGTTTCTTAAATTGA[G/T]ATCCCTTGAAGGATGATAGGAAGTA
- Phenotype
-
MIM: 601235
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DDX10
PubMed Links
Gene Details
- Gene
- DDX10
- Gene Name
- DEAD-box helicase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004398.3 |
|
Intron |
|
|
NP_004389.2 |
View Full Product Details