Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_032449.2 | 6549 | Intron | NP_115825.1 | ||
XM_005270590.2 | 6549 | UTR 3 | XP_005270647.1 | ||
XM_005270591.2 | 6549 | Intron | XP_005270648.1 | ||
XM_005270593.2 | 6549 | Intron | XP_005270650.1 | ||
XM_006710423.2 | 6549 | Intron | XP_006710486.1 | ||
XM_017000559.1 | 6549 | Intron | XP_016856048.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004799.3 | 6549 | Intron | NP_004790.2 | ||
NM_007324.3 | 6549 | Intron | NP_015563.2 | ||
XM_011542437.2 | 6549 | Intron | XP_011540739.1 | ||
XM_017002845.1 | 6549 | Intron | XP_016858334.1 |