Product Details

SNP ID
rs35080429
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10573773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACCAAGTGAACAATGGGGCCAG[C/G]TGAGCCTGGCCCTGCTTTAAGGGAG
Phenotype
MIM: 605702 MIM: 604673
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LYVE1 PubMed Links
Additional Information
For this assay, SNP(s) [rs75750657] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LYVE1
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
There are no transcripts associated with this gene.

Gene
MRVI1
Gene Name
murine retrovirus integration site 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098579.2 5631 UTR 3 NP_001092049.2
NM_001100163.2 5631 UTR 3 NP_001093633.1
NM_001100167.2 5631 UTR 3 NP_001093637.1
NM_001206880.1 5631 UTR 3 NP_001193809.1
NM_001206881.1 5631 UTR 3 NP_001193810.1
NM_130385.3 5631 UTR 3 NP_569056.4
Gene
MRVI1-AS1
Gene Name
MRVI1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details