Product Details

SNP ID
rs13196459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2888837 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAGTTAATTTTATGTTATGTGA[A/C]GTTTACCTCAATAAAAAAATTCTTA
Phenotype
MIM: 601799
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927730 PubMed Links

Gene Details

Gene
LOC101927730
Gene Name
uncharacterized LOC101927730
There are no transcripts associated with this gene.

Gene
SERPINB9
Gene Name
serpin family B member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004155.5 2729 UTR 3 NP_004146.1
XM_005249184.4 2729 UTR 3 XP_005249241.1
XM_011514678.2 2729 UTR 3 XP_011512980.1
XM_017010942.1 2729 UTR 3 XP_016866431.1
XM_017010943.1 2729 UTR 3 XP_016866432.1

View Full Product Details