Product Details

SNP ID
rs2328510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20394344 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCCTCCCTTCTTCTAGGCTCAGT[A/C]AGGTCGGGCATACAGAGAAGGCTCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RALGAPA2 PubMed Links

Gene Details

Gene
RALGAPA2
Gene Name
Ral GTPase activating protein catalytic alpha subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020343.3 Intron NP_065076.2
XM_005260768.2 Intron XP_005260825.1
XM_006723598.1 Intron XP_006723661.1
XM_006723599.1 Intron XP_006723662.1
XM_011529309.1 Intron XP_011527611.1
XM_011529310.2 Intron XP_011527612.1
XM_011529311.2 Intron XP_011527613.2
XM_017027972.1 Intron XP_016883461.1
XM_017027973.1 Intron XP_016883462.1
XM_017027974.1 Intron XP_016883463.1
XM_017027975.1 Intron XP_016883464.1
XM_017027976.1 Intron XP_016883465.1
XM_017027977.1 Intron XP_016883466.1

View Full Product Details