Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001024688.2 | 2751 | UTR 3 | NP_001019859.1 | ||
NM_002485.4 | 2751 | UTR 3 | NP_002476.2 | ||
XM_011517045.1 | 2751 | UTR 3 | XP_011515347.1 | ||
XM_011517046.1 | 2751 | Intron | XP_011515348.1 | ||
XM_017013460.1 | 2751 | UTR 3 | XP_016868949.1 | ||
XM_017013461.1 | 2751 | UTR 3 | XP_016868950.1 | ||
XM_017013462.1 | 2751 | UTR 3 | XP_016868951.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001126111.2 | 2751 | Intron | NP_001119583.1 | ||
NM_004337.2 | 2751 | Intron | NP_004328.1 |