Product Details

SNP ID
rs2331291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26662857 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAAGAACAGTACATGCTGTTTAG[C/T]GGGAAAAATGAATATCTCATGGGTA
Phenotype
MIM: 611082
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIAT PubMed Links

Gene Details

Gene
MIAT
Gene Name
myocardial infarction associated transcript (non-protein coding)
There are no transcripts associated with this gene.

Gene
MIATNB
Gene Name
MIAT neighbor (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details