Product Details

SNP ID
rs232262
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:45722292 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCTGGCCCTTGGGGTAGGTGAGCT[C/G]AACCGTCTCAAGTATCAAGCCCAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPINT4 PubMed Links

Gene Details

Gene
SPINT4
Gene Name
serine peptidase inhibitor, Kunitz type 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178455.1 Intron NP_848550.1
Gene
WFDC3
Gene Name
WAP four-disulfide core domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080614.1 Intron NP_542181.1
XM_011528553.2 Intron XP_011526855.1
XM_017027668.1 Intron XP_016883157.1

View Full Product Details