Product Details

SNP ID
rs6950541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:15615467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGAGTATGTGGACGCCATCCAT[A/G]TGATTACTATCATTCAGGTAATTCT
Phenotype
MIM: 600535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEOX2 PubMed Links

Gene Details

Gene
MEOX2
Gene Name
mesenchyme homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005924.4 Intron NP_005915.2

View Full Product Details