Product Details
- SNP ID
-
rs6461197
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:15616049 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTCTATGTCTATCGTTGAAATT[G/T]TACCATTTTATTTTTTAAAATAGAT
- Phenotype
-
MIM: 600535
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MEOX2
PubMed Links
Gene Details
- Gene
- MEOX2
- Gene Name
- mesenchyme homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005924.4 |
|
Intron |
|
|
NP_005915.2 |
View Full Product Details