Product Details

SNP ID
rs4714025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36948323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGATGACCACACCAGGGCAACCT[A/G]TAGTGCCAGTGCCTGGTTCCACCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PI16 PubMed Links

Gene Details

Gene
PI16
Gene Name
peptidase inhibitor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199159.1 61 UTR 5 NP_001186088.1
NM_153370.2 61 Intron NP_699201.2
XM_005248917.2 61 Intron XP_005248974.1
XM_011514375.2 61 Intron XP_011512677.1
XM_017010430.1 61 Intron XP_016865919.1

View Full Product Details