Product Details

SNP ID
rs10762242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68832373 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCAGGGCCTTTCATCTTTTCTCA[C/T]TGAATAAAAATGCCCCAGGCCATGC
Phenotype
MIM: 609397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STOX1 PubMed Links

Gene Details

Gene
STOX1
Gene Name
storkhead box 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130159.2 Intron NP_001123631.1
NM_001130160.2 Intron NP_001123632.1
NM_001130161.2 Intron NP_001123633.1
NM_152709.4 Intron NP_689922.3
XM_011539454.2 Intron XP_011537756.1

View Full Product Details