Product Details

SNP ID
rs12011827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:147990884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACATGGGACTAACCTTCTTCACC[A/G]CACCGCTGAAACCACTAACCTCCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FMR1NB PubMed Links

Gene Details

Gene
FMR1NB
Gene Name
fragile X mental retardation 1 neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152578.2 Intron NP_689791.1

View Full Product Details