Product Details

SNP ID
rs11651212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48623779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAGAGCCACAAACAGCCAGCACA[A/C]TCCAATTCACACGCAATGCAATTGC
Phenotype
MIM: 142963 MIM: 142964 MIM: 608632
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HOXB8 PubMed Links

Gene Details

Gene
HOXB8
Gene Name
homeobox B8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024016.3 Intron NP_076921.1
XM_005257286.2 Intron XP_005257343.1
XM_017024564.1 Intron XP_016880053.1
Gene
HOXB9
Gene Name
homeobox B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024017.4 Intron NP_076922.1
Gene
MIR196A1
Gene Name
microRNA 196a-1
There are no transcripts associated with this gene.

View Full Product Details