Product Details

SNP ID
rs11757602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31465025 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATGAGTTACACAGATGCTAATTC[G/T]TAGGGGCACTTGGAATCACATGGTT
Phenotype
MIM: 604676
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HCG26 PubMed Links

Gene Details

Gene
HCG26
Gene Name
HLA complex group 26 (non-protein coding)
There are no transcripts associated with this gene.

Gene
HCP5
Gene Name
HLA complex P5 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details