Product Details

SNP ID
rs11134652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170785123 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAATAGCCACTTAACGATTGGCTC[A/G]TCTACCACAGGGAGTGATGGCATCA
Phenotype
MIM: 602729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRP PubMed Links

Gene Details

Gene
GABRP
Gene Name
gamma-aminobutyric acid type A receptor pi subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291985.1 Intron NP_001278914.1
NM_014211.2 Intron NP_055026.1
XM_005265872.2 Intron XP_005265929.1

View Full Product Details