Product Details

SNP ID
rs11829948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54000553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCACTACGCCCTCAAGCCGGACG[C/T]CTACCCCGGGCGCCGCGCGGACTGC
Phenotype
MIM: 142970 MIM: 142971 MIM: 609687
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXC-AS1 PubMed Links

Gene Details

Gene
HOXC-AS1
Gene Name
HOXC cluster antisense RNA 1
There are no transcripts associated with this gene.

Gene
HOXC-AS2
Gene Name
HOXC cluster antisense RNA 2
There are no transcripts associated with this gene.

Gene
HOXC8
Gene Name
homeobox C8
There are no transcripts associated with this gene.

Gene
HOXC9
Gene Name
homeobox C9
There are no transcripts associated with this gene.

Gene
LOC107984512
Gene Name
uncharacterized LOC107984512
There are no transcripts associated with this gene.

Gene
MIR196A2
Gene Name
microRNA 196a-2
There are no transcripts associated with this gene.

View Full Product Details