Product Details

SNP ID
rs11988213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66669985 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAACACCAGGCCAAATTTTTCTTA[C/T]TGATGATACATATACATAGTGAAGT
Phenotype
MIM: 611745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf44 PubMed Links

Gene Details

Gene
C8orf44
Gene Name
chromosome 8 open reading frame 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019607.2 Intron NP_062553.1
Gene
C8orf44-SGK3
Gene Name
C8orf44-SGK3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204173.1 Intron NP_001191102.1
Gene
VCPIP1
Gene Name
valosin containing protein interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details