Product Details

SNP ID
rs1802115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17247099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGCTCAGGATGGATGTTAGTTT[A/C]AAGAATAACTGGATGGTTTGTTTCC
Phenotype
MIM: 604665 MIM: 607273
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COPS3 PubMed Links

Gene Details

Gene
COPS3
Gene Name
COP9 signalosome subunit 3
There are no transcripts associated with this gene.

Gene
FLCN
Gene Name
folliculin
There are no transcripts associated with this gene.

View Full Product Details