Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199125.1 | Intron | NP_001186054.1 | |||
NM_001316354.1 | Intron | NP_001303283.1 | |||
NM_001316355.1 | Intron | NP_001303284.1 | |||
NM_001316356.1 | Intron | NP_001303285.1 | |||
NM_001316357.1 | Intron | NP_001303286.1 | |||
NM_001316358.1 | Intron | NP_001303287.1 | |||
NM_003653.3 | Intron | NP_003644.2 | |||
XM_005256837.4 | Intron | XP_005256894.1 | |||
XM_005256840.3 | Intron | XP_005256897.1 | |||
XM_005256842.4 | Intron | XP_005256899.1 | |||
XM_017025246.1 | Intron | XP_016880735.1 |