Product Details

SNP ID
rs1413974
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:23020632 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTTGACTCCAAACAGTATATACAGA[C/T]AGTGACTGCCTCAAATGTCTGTGTC
Phenotype
MIM: 609132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf234 PubMed Links

Gene Details

Gene
C1orf234
Gene Name
chromosome 1 open reading frame 234
There are no transcripts associated with this gene.

Gene
KDM1A
Gene Name
lysine demethylase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009999.2 Intron NP_001009999.1
NM_015013.3 Intron NP_055828.2
XM_005245786.2 Intron XP_005245843.1
XM_006710472.3 Intron XP_006710535.1
XM_006710473.2 Intron XP_006710536.1
XM_006710474.3 Intron XP_006710537.1
XM_017000716.1 Intron XP_016856205.1
XM_017000717.1 Intron XP_016856206.1
XM_017000718.1 Intron XP_016856207.1
XM_017000719.1 Intron XP_016856208.1
XM_017000720.1 Intron XP_016856209.1

View Full Product Details