Product Details

SNP ID
hCV7609043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102293131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGGGGCGGCGAGTCCGACTCAT[A/C]AATATTTTAAGGAATGACCCGGCCC
Phenotype
MIM: 116896 MIM: 605300
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CUX1 PubMed Links

Gene Details

Gene
CUX1
Gene Name
cut like homeobox 1
There are no transcripts associated with this gene.

Gene
MIR4285
Gene Name
microRNA 4285
There are no transcripts associated with this gene.

Gene
SH2B2
Gene Name
SH2B adaptor protein 2
There are no transcripts associated with this gene.

View Full Product Details