Product Details

SNP ID
rs11625621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:51246941 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAATTGGGGGCTAAAATGTCATGA[C/G]TAGGTAAATAGTATAATTTGCTATT
Phenotype
MIM: 610527
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMX1 PubMed Links

Gene Details

Gene
TMX1
Gene Name
thioredoxin related transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030755.4 Intron NP_110382.3

View Full Product Details