Product Details

SNP ID
rs1049749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49187899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTTCTTTGAACAATACCGAAGG[T/G]TTTGGGAAAGGGTAGGGGTCTGGAG
Phenotype
MIM: 300111 MIM: 313475
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
PRICKLE3 PubMed Links

Gene Details

Gene
PRICKLE3
Gene Name
prickle planar cell polarity protein 3
There are no transcripts associated with this gene.

Gene
SYP
Gene Name
synaptophysin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003179.2 2346 UTR 3 NP_003170.1

View Full Product Details