Product Details

SNP ID
rs1513890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75914211 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCACAAACTCCAAATCTCCTGGAC[A/C]CACTTCGCAAGGTTGTAAAGTTAAA
Phenotype
MIM: 607469
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAAA PubMed Links

Gene Details

Gene
NAAA
Gene Name
N-acylethanolamine acid amidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042402.1 1237 Intron NP_001035861.1
NM_014435.3 1237 UTR 3 NP_055250.2
XM_005262920.3 1237 Intron XP_005262977.1
XM_005262921.3 1237 Intron XP_005262978.1
XM_005262923.2 1237 UTR 3 XP_005262980.1
XM_005262924.2 1237 UTR 3 XP_005262981.1
XM_005262925.3 1237 Intron XP_005262982.1
XM_006714180.2 1237 Intron XP_006714243.1
XM_011531852.2 1237 Intron XP_011530154.1
XM_011531854.2 1237 Intron XP_011530156.1
XM_017008027.1 1237 Intron XP_016863516.1
XM_017008028.1 1237 UTR 3 XP_016863517.1
XM_017008029.1 1237 Intron XP_016863518.1
XM_017008030.1 1237 UTR 3 XP_016863519.1

View Full Product Details