Product Details

SNP ID
rs11413
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:35178261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATTAGAATGAAAACTGTATTTT[C/T]TTTAAAATGTAAAAGCCTGACTCTC
Phenotype
MIM: 605199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFPQ PubMed Links

Gene Details

Gene
SFPQ
Gene Name
splicing factor proline and glutamine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005066.2 8431 Intron NP_005057.1
XM_005271112.4 8431 Intron XP_005271169.1
XM_005271113.4 8431 Intron XP_005271170.1
XM_005271115.4 8431 Intron XP_005271172.1
XM_011541950.2 8431 UTR 3 XP_011540252.1
XM_017002053.1 8431 UTR 3 XP_016857542.1
XM_017002054.1 8431 UTR 3 XP_016857543.1

View Full Product Details