Product Details
- SNP ID
-
rs1047429
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:3897606 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGAGCGGCTGAGGCTAGGGTAGG[C/G]CTCCCTCAGGGTGTGTGGGGTCGGA
- Phenotype
-
MIM: 611411
MIM: 600845
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CAMKK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117126856] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAMKK1
- Gene Name
- calcium/calmodulin dependent protein kinase kinase 1
There are no transcripts associated with this gene.
- Gene
- P2RX1
- Gene Name
- purinergic receptor P2X 1
View Full Product Details