Product Details

SNP ID
rs908124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101401580 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCGGATAGCGGCTAAACATCAAA[C/T]ATGTGCAGAGAAATTAGCCCTAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RFX8 PubMed Links

Gene Details

Gene
RFX8
Gene Name
RFX family member 8, lacking RFX DNA binding domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145664.1 Intron NP_001139136.1
XM_011511771.2 Intron XP_011510073.1
XM_011511772.1 Intron XP_011510074.1
XM_011511773.1 Intron XP_011510075.1
XM_011511776.1 Intron XP_011510078.1
XM_011511777.1 Intron XP_011510079.1
XM_011511778.1 Intron XP_011510080.1
XM_017004851.1 Intron XP_016860340.1
XM_017004852.1 Intron XP_016860341.1
XM_017004853.1 Intron XP_016860342.1
XM_017004854.1 Intron XP_016860343.1

View Full Product Details