Product Details

SNP ID
hCV8771416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:129832785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGTAATTAGAACACTATTGTTT[A/T]TACAATATTGAAAAAATACAATTTT
Phenotype
MIM: 601082
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2H PubMed Links

Gene Details

Gene
UBE2H
Gene Name
ubiquitin conjugating enzyme E2 H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202498.1 3016 UTR 3 NP_001189427.1
NM_003344.3 3016 UTR 3 NP_003335.1
NM_182697.2 3016 UTR 3 NP_874356.1

View Full Product Details