Product Details

SNP ID
rs1061479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71750712 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACTGCTGGTAGGAGGCAGCTGGG[G/T]AAGGTGGCGATGGCCAGTGGGCAGG
Phenotype
MIM: 615608 MIM: 605516
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C10orf54 PubMed Links

Gene Details

Gene
C10orf54
Gene Name
chromosome 10 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022153.1 1618 UTR 3 NP_071436.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 1618 Intron NP_001165401.1
NM_001171931.1 1618 Intron NP_001165402.1
NM_001171932.1 1618 Intron NP_001165403.1
NM_001171933.1 1618 Intron NP_001165404.1
NM_001171934.1 1618 Intron NP_001165405.1
NM_001171935.1 1618 Intron NP_001165406.1
NM_001171936.1 1618 Intron NP_001165407.1
NM_022124.5 1618 Intron NP_071407.4
NM_052836.3 1618 Intron NP_443068.1
XM_006717940.3 1618 Intron XP_006718003.1
XM_006717942.3 1618 Intron XP_006718005.1
XM_011540039.2 1618 Intron XP_011538341.1
XM_011540042.2 1618 Intron XP_011538344.1
XM_011540043.2 1618 Intron XP_011538345.1
XM_011540044.2 1618 Intron XP_011538346.1
XM_011540045.2 1618 Intron XP_011538347.1
XM_011540046.2 1618 Intron XP_011538348.1
XM_011540047.2 1618 Intron XP_011538349.1
XM_011540048.2 1618 Intron XP_011538350.1
XM_011540049.2 1618 Intron XP_011538351.1
XM_011540051.2 1618 Intron XP_011538353.1
XM_011540052.2 1618 Intron XP_011538354.1
XM_017016499.1 1618 Intron XP_016871988.1
XM_017016500.1 1618 Intron XP_016871989.1
XM_017016501.1 1618 Intron XP_016871990.1
XM_017016502.1 1618 Intron XP_016871991.1
XM_017016503.1 1618 Intron XP_016871992.1
XM_017016504.1 1618 Intron XP_016871993.1
XM_017016505.1 1618 Intron XP_016871994.1
XM_017016506.1 1618 Intron XP_016871995.1
XM_017016507.1 1618 Intron XP_016871996.1
XM_017016508.1 1618 Intron XP_016871997.1

View Full Product Details