Product Details

SNP ID
rs369887
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:55840801 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGTGGTTATGCAGCATCTAGATTT[C/G]TCACCAATGTTTTGGCTTGGGGTCT
Phenotype
MIM: 609664 MIM: 609645
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NLRP11 PubMed Links

Gene Details

Gene
NLRP11
Gene Name
NLR family pyrin domain containing 11
There are no transcripts associated with this gene.

Gene
NLRP4
Gene Name
NLR family pyrin domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_134444.4 Intron NP_604393.2
XM_017026344.1 Intron XP_016881833.1
XM_017026345.1 Intron XP_016881834.1

View Full Product Details