Product Details

SNP ID
rs1051177
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:74705866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGTTTAGAATAGAGGTTTTTAG[A/T]TCAGAAAATGTTCAAAGTCCAATAC
Phenotype
MIM: 123691
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CRYZ PubMed Links

Gene Details

Gene
CRYZ
Gene Name
crystallin zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130042.1 1409 UTR 3 NP_001123514.1
NM_001130043.1 1409 UTR 3 NP_001123515.1
NM_001134759.1 1409 UTR 3 NP_001128231.1
NM_001889.3 1409 UTR 3 NP_001880.2
XM_005270491.4 1409 UTR 3 XP_005270548.1
XM_011540747.1 1409 UTR 3 XP_011539049.1
XM_017000367.1 1409 UTR 3 XP_016855856.1
XM_017000368.1 1409 UTR 3 XP_016855857.1

View Full Product Details