Product Details

SNP ID
rs1365494
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:99885095 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGACATGCAAAAACTAAAAACTAA[C/T]TTCTTTCCCACAAATAATAATCTCA
Phenotype
MIM: 603296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMTOR3 PubMed Links

Gene Details

Gene
LAMTOR3
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243736.1 Intron NP_001230665.1
NM_021970.3 Intron NP_068805.1

View Full Product Details