Product Details
- SNP ID
-
rs4867841
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:176649233 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGATGGTGAGGTGGGAAAGGTGAGG[G/A]AGGTTGCTGGGAAAGTCAAGAGGCT
- Phenotype
-
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EIF4E1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116167787] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EIF4E1B
- Gene Name
- eukaryotic translation initiation factor 4E family member 1B
There are no transcripts associated with this gene.
- Gene
- TSPAN17
- Gene Name
- tetraspanin 17
View Full Product Details