Product Details

SNP ID
rs57117264
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:176648393 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGCTCCTGGGCGGGTCTGCCCCTAC[C/T]GGGGAAGGGAGCTACTAGTGCCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF4E1B PubMed Links

Gene Details

Gene
EIF4E1B
Gene Name
eukaryotic translation initiation factor 4E family member 1B
There are no transcripts associated with this gene.

Gene
TSPAN17
Gene Name
tetraspanin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006616.2 Intron NP_001006617.2
NM_012171.2 Intron NP_036303.1
NM_130465.4 Intron NP_569732.2
XM_005265876.3 Intron XP_005265933.1

View Full Product Details