Product Details

SNP ID
rs1204508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:430875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATTATTTTTGTAGAGATGAAGT[A/G]TAGTATAGTGATACTGCCCAGGCAT
Phenotype
MIM: 608738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB11FIP3 PubMed Links

Gene Details

Gene
RAB11FIP3
Gene Name
RAB11 family interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142272.1 Intron NP_001135744.1
NM_014700.3 Intron NP_055515.1
XM_005255713.3 Intron XP_005255770.1
XM_005255714.1 Intron XP_005255771.1
XM_005255715.4 Intron XP_005255772.1
XM_005255717.3 Intron XP_005255774.1
XM_005255718.3 Intron XP_005255775.1
XM_011522764.2 Intron XP_011521066.1
XM_011522765.2 Intron XP_011521067.1
XM_017023907.1 Intron XP_016879396.1

View Full Product Details