Product Details

SNP ID
rs1048005
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:64834316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCACAAGGAGGAGTGCTTAAGTG[C/G]TTTCAAATATTTTGGTTGTCATTAT
Phenotype
MIM: 147795 MIM: 609953
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
JAK1 PubMed Links

Gene Details

Gene
JAK1
Gene Name
Janus kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320923.1 4022 UTR 3 NP_001307852.1
NM_001321852.1 4022 UTR 3 NP_001308781.1
NM_001321853.1 4022 UTR 3 NP_001308782.1
NM_001321854.1 4022 UTR 3 NP_001308783.1
NM_001321855.1 4022 UTR 3 NP_001308784.1
NM_001321856.1 4022 UTR 3 NP_001308785.1
NM_001321857.1 4022 UTR 3 NP_001308786.1
NM_002227.3 4022 UTR 3 NP_002218.2
XM_017001212.1 4022 UTR 3 XP_016856701.1
Gene
RAVER2
Gene Name
ribonucleoprotein, PTB binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018211.3 4022 Intron NP_060681.2

View Full Product Details