Product Details

SNP ID
rs977708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32740880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTTGCCAAGATGGTGCTGTGCAG[C/T]CTTCACTGCTTCTGGGACCACACGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TDRD12 PubMed Links

Gene Details

Gene
TDRD12
Gene Name
tudor domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110822.1 Intron NP_001104292.1
XM_011527471.2 Intron XP_011525773.1
XM_011527472.2 Intron XP_011525774.1
XM_011527473.2 Intron XP_011525775.1
XM_011527474.2 Intron XP_011525776.1
XM_011527475.2 Intron XP_011525777.1
XM_011527476.2 Intron XP_011525778.1
XM_017027458.1 Intron XP_016882947.1
XM_017027459.1 Intron XP_016882948.1

View Full Product Details