Product Details
- SNP ID
-
rs10766785
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:3222602 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGGGGCACGGGTGTCGGCCCCGC[A/C]GAGGCTGGGCTTTGGTCTGTCTTGT
- Phenotype
-
MIM: 607232
MIM: 607234
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MRGPRE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115984363] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRGPRE
- Gene Name
- MAS related GPR family member E
There are no transcripts associated with this gene.
- Gene
- MRGPRG
- Gene Name
- MAS related GPR family member G
There are no transcripts associated with this gene.
- Gene
- MRGPRG-AS1
- Gene Name
- MRGPRG antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details