Product Details

SNP ID
rs3820678
Assay Type
Functionally tested
NCBI dbSNP Submissions
83
Location
Chr.1:158480448 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGTTTTCAGCCTCCCTTTTTGTAGC[A/G]CCAACAAAGTCAATCATTACTTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10R2 PubMed Links

Gene Details

Gene
OR10R2
Gene Name
olfactory receptor family 10 subfamily R member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004472.1 571 Missense Mutation ACC,GCC T191A NP_001004472.1

View Full Product Details